Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARA7
Entrez Gene ID
827706
Gene Name
Ras-related small GTP-binding family protein
Gene Aliases
ARA-7, ARABIDOPSIS RAB GTPASE HOMOLOG F2B, AT4G19640, ATRAB-F2B, ATRAB5B, ATRABF2B, F24J7.190, F24J7_190, RAB GTPASE HOMOLOG F2B, RAB-F2B, RABF2B
Chromosome Location
Chr.4: 10687196 - 10689748 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001341360.1 NP_001327978.1 6 - 7 876 123
NM_001341361.1 NP_001327977.1 6 - 7 1396 123
NM_118084.5 NP_193699.1 6 - 7 801 123
GenBank mRNA AB038491.1 - 6 - 7 502 123
AF370309.1 - 6 - 7 628 123
AY052670.1 - 6 - 7 664 123
AY063095.1 - 6 - 7 502 123

View Full Product Details