Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- NRPB2
- Entrez Gene ID
-
828259
- Gene Name
- DNA-directed RNA polymerase family protein
- Gene Aliases
-
AT4G21710, EMB1989, EMBRYO DEFECTIVE 1989, F17L22.170, F17L22_170, RPB2
- Chromosome Location
-
Chr.4: 11535404 - 11542405 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_118291.4 |
NP_193902.1 |
23 - 24 |
3240 |
78 |
View Full Product Details