Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ROC1
Entrez Gene ID
830029
Gene Name
rotamase CYP 1
Gene Aliases
AT4G38740, CYCLOPHILIN, T9A14.20, T9A14_20, rotamase CYP 1
Chromosome Location
Chr.4: 18083395 - 18084630 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120034.2 NP_195585.1 1 - 1 780 139
GenBank mRNA AY093227.1 - 1 - 1 724 139
L14844.1 - 1 - 1 716 139

View Full Product Details