Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NIR1
Entrez Gene ID
816055
Gene Name
nitrite reductase 1
Gene Aliases
ARABIDOPSIS THALIANA NITRITE REDUCTASE, AT2G15620, ATHNIR, NIR, NITRITE REDUCTASE, nitrite reductase 1
Chromosome Location
Chr.2: 6810395 - 6812941 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127123.3 NP_179164.1 1 - 2 533 81
GenBank mRNA AB006032.1 - 1 - 2 387 81
AF360320.1 - 1 - 2 400 81
AY045608.1 - 1 - 2 398 81
AY093995.1 - 1 - 2 376 81
AY142685.1 - 1 - 2 376 81

View Full Product Details