Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRMT4B
Entrez Gene ID
819878
Gene Name
protein arginine methyltransferase 4B
Gene Aliases
ARABIDOPSIS THALIANA PROTEIN ARGININE METHYLTRANSFERASE 4B, AT3G06930, ATPRMT4B, protein arginine methyltransferase 4B
Chromosome Location
Chr.3: 2185129 - 2189434 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111573.5 NP_187349.2 3 - 4 495 109
NM_180197.2 NP_850528.1 3 - 4 448 109
GenBank mRNA BT015414.1 - 3 - 4 357 109

View Full Product Details