Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HXK2
Entrez Gene ID
816505
Gene Name
hexokinase 2
Gene Aliases
ARABIDOPSIS THALIANA HEXOKINASE 2, AT2G19860, ATHXK2, F6F22.11, F6F22_11, HEXOKINASE 2, hexokinase 2
Chromosome Location
Chr.2: 8570607 - 8574067 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084452.2 NP_001077921.1 2 - 3 790 82
NM_127544.3 NP_179576.1 3 - 4 821 82

View Full Product Details