Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RTFL16
Entrez Gene ID
822160
Gene Name
ROTUNDIFOLIA like 16
Gene Aliases
AT3G25717, DEVIL 6, DVL6, ROTUNDIFOLIA like 16
Chromosome Location
Chr.3: 9375102 - 9375671 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_180300.1 NP_850631.2 1 - 1 285 146

View Full Product Details