Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RBL4
Entrez Gene ID
824545
Gene Name
RHOMBOID-like protein 4
Gene Aliases
AT3G53780, ATRBL4, RHOMBOID-like protein 4
Chromosome Location
Chr.3: 19924538 - 19927026 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339632.1 NP_001325690.1 2 - 3 905 137
NM_115238.2 NP_566989.1 3 - 4 835 137
NM_180367.2 NP_850698.1 2 - 3 905 137
GenBank mRNA BT029533.1 - 1 - 2 155 137

View Full Product Details