The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001036309.2 | NP_001031386.1 | 4 - 5 | 742 | 104 |
NM_001036310.1 | NP_001031387.1 | 4 - 5 | 714 | 104 | |
NM_127705.4 | NP_565508.1 | 4 - 5 | 1121 | 104 | |
GenBank mRNA | AF419589.1 | - | 4 - 5 | 710 | 104 |
AF428408.1 | - | 4 - 5 | 710 | 104 | |
AY049282.1 | - | 4 - 5 | 710 | 104 | |
AY049286.1 | - | 4 - 5 | 710 | 104 | |
AY062582.1 | - | 4 - 5 | 710 | 104 | |
AY090291.1 | - | 4 - 5 | 654 | 104 | |
AY128380.1 | - | 4 - 5 | 712 | 104 | |
AY128784.1 | - | 4 - 5 | 654 | 104 | |
AY142633.1 | - | 4 - 5 | 654 | 104 | |
BT000106.1 | - | 4 - 5 | 654 | 104 |