Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RNR1
Entrez Gene ID
816715
Gene Name
ribonucleotide reductase 1
Gene Aliases
AT2G21790, ATRNR1, CLS8, CRINKLY LEAVES 8, DPD2, R1, RIBONUCLEOTIDE REDUCTASE 1, RIBONUCLEOTIDE REDUCTASE LARGE SUBUNIT 1, defective in pollen organelle DNA degradation 2, ribonucleotide reductase 1
Chromosome Location
Chr.2: 9293261 - 9297861 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127748.4 NP_179770.1 3 - 4 642 73
GenBank mRNA AY035080.1 - 3 - 4 548 73
BT008573.1 - 3 - 4 374 73

View Full Product Details