Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IAA9
Entrez Gene ID
836693
Gene Name
indole-3-acetic acid inducible 9
Gene Aliases
AT5G65670, INDOLE-3-ACETIC ACID 9, MPA24.1, MPA24_1, indole-3-acetic acid inducible 9
Chromosome Location
Chr.5: 26253408 - 26256544 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001345703.1 NP_001318885.1 2 - 3 771 77
NM_180944.3 NP_851275.1 2 - 3 1066 77
GenBank mRNA AF334715.1 - 1 - 2 543 77
U18411.1 - 2 - 3 776 77

View Full Product Details