Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WOX4
Entrez Gene ID
841113
Gene Name
WUSCHEL related homeobox 4
Gene Aliases
AT1G46480, F2G19.11, F2G19_11, WUSCHEL related homeobox 4
Chromosome Location
Chr.1: 17236524 - 17238185 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001333264.1 NP_001320246.1 1 - 2 476 83
NM_103605.7 NP_175145.2 1 - 2 476 83
GenBank mRNA AB493501.1 - 1 - 2 244 83
AY251396.1 - 1 - 2 244 83
BT010692.1 - 1 - 2 344 83
BT010974.1 - 1 - 2 244 83
FJ440850.1 - 1 - 2 277 83

View Full Product Details