Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RBL15
Entrez Gene ID
825015
Gene Name
RHOMBOID-like protein 15
Gene Aliases
ARABIDOPSIS RHOMBOID-LIKE PROTEIN 11, AT3G58460, ATRBL11, ATRBL15, RBL11, RHOMBOID-like protein 15, rhomboid-like protein 11
Chromosome Location
Chr.3: 21623179 - 21626701 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203197.1 NP_001190126.1 4 - 5 626 115
NM_115708.4 NP_567064.1 3 - 4 759 115
GenBank mRNA AY136386.1 - 3 - 4 593 115
BT000186.1 - 3 - 4 498 115

View Full Product Details