Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MYB17
Entrez Gene ID
825297
Gene Name
myb domain protein 17
Gene Aliases
AT3G61250, AtMYB17, LATE MERISTEM IDENTITY2, LMI2, myb domain protein 17
Chromosome Location
Chr.3: 22670729 - 22672751 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115989.4 NP_191684.1 1 - 2 711 74
GenBank mRNA AY519601.1 - 1 - 2 134 74
BT033129.1 - 1 - 2 134 74

View Full Product Details