Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VSR2
Entrez Gene ID
817579
Gene Name
VACUOLAR SORTING RECEPTOR 2
Gene Aliases
AT2G30290, AtVSR2, BP80-1;2, T9D9.10, T9D9_10, VACUOLAR SORTING RECEPTOR 1;2, VACUOLAR SORTING RECEPTOR 2, VSR1;2, binding protein of 80 kDa 1;2
Chromosome Location
Chr.2: 12912519 - 12915915 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202708.1 NP_001189637.1 6 - 7 1368 76
NM_128582.2 NP_180588.1 6 - 7 1371 76
GenBank mRNA DQ446580.1 - 6 - 7 1237 76

View Full Product Details