Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WRKY72
Entrez Gene ID
831365
Gene Name
WRKY DNA-binding protein 72
Gene Aliases
ARABIDOPSIS THALIANA WRKY DNA-BINDING PROTEIN 72, AT5G15130, ATWRKY72, F8M21.20, F8M21_20, WRKY DNA-binding protein 72
Chromosome Location
Chr.5: 4904290 - 4907094 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001343389.1 NP_001331284.1 1 - 2 619 129
NM_121517.3 NP_197017.1 2 - 3 808 129

View Full Product Details