Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SYN4
- Entrez Gene ID
-
831487
- Gene Name
- sister chromatid cohesion 1 protein 4
- Gene Aliases
-
ARABIDOPSIS HOMOLOG OF RAD21 3, AT5G16270, ATRAD21.3, T21H19.190, T21H19_190, sister chromatid cohesion 1 protein 4
- Chromosome Location
-
Chr.5: 5316197 - 5322700 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_121632.4 |
NP_197131.2 |
11 - 12 |
2182 |
76 |
GenBank mRNA |
AF400129.1 |
- |
11 - 12 |
2080 |
76 |
View Full Product Details