Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ENH1
Entrez Gene ID
831581
Gene Name
rubredoxin family protein
Gene Aliases
AT5G17170, MKP11.2, MKP11_2, enhancer of sos3-1
Chromosome Location
Chr.5: 5649057 - 5651205 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085129.1 NP_001078598.1 2 - 3 470 110
NM_121723.5 NP_568342.1 2 - 3 606 110
GenBank mRNA AY035119.1 - 2 - 3 413 110
BT000984.1 - 2 - 3 328 110

View Full Product Details