Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CSA1
Entrez Gene ID
831656
Gene Name
disease resistance protein (TIR-NBS-LRR class)
Gene Aliases
AT5G17880, MPI7.20, MPI7_20, constitutive shade-avoidance1
Chromosome Location
Chr.5: 5908834 - 5913317 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001343529.1 NP_001331999.1 1 - 2 701 99
NM_121794.2 NP_197290.1 1 - 2 483 99

View Full Product Details