Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NCBP
Entrez Gene ID
831929
Gene Name
Putative cap-binding protein
Gene Aliases
AT5G18110, MRG7.7, MRG7_7, novel cap-binding protein
Chromosome Location
Chr.5: 5988744 - 5990681 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121816.2 NP_197312.1 5 - 6 628 91
GenBank mRNA AY092964.1 - 5 - 6 628 91
AY093208.1 - 5 - 6 624 91
AY114562.1 - 5 - 6 535 91

View Full Product Details