Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ZIP4
- Entrez Gene ID
-
842113
- Gene Name
- Clathrin adaptor complexes medium subunit family protein
- Gene Aliases
-
AP3M, AT1G56590, F25P12.96, F25P12_96, ZIG SUPPRESSOR 4, adaptor protein-3 mu-adaptin
- Chromosome Location
-
Chr.1: 21201948 - 21204856 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_104536.5 |
NP_176052.3 |
6 - 7 |
951 |
128 |
View Full Product Details