Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GPX8
Entrez Gene ID
842652
Gene Name
glutathione peroxidase 8
Gene Aliases
AT1G63460, ATGPX8, F2K11.16, F2K11_16, glutathione peroxidase 8
Chromosome Location
Chr.1: 23534946 - 23536467 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105025.4 NP_564813.1 1 - 2 212 121
GenBank mRNA AK227611.1 - 1 - 2 142 121
AY086991.1 - 1 - 2 56 121
BT003059.1 - 1 - 2 40 121

View Full Product Details