Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CP5
- Entrez Gene ID
-
842780
- Gene Name
- Polyketide cyclase/dehydrase and lipid transport superfamily protein
- Gene Aliases
-
AT1G64720, F13O11.4, F13O11_4
- Chromosome Location
-
Chr.1: 24046553 - 24048822 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_105147.4 |
NP_176653.1 |
5 - 6 |
1024 |
73 |
View Full Product Details