Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAPR4
Entrez Gene ID
827155
Gene Name
membrane-associated progesterone binding protein 4
Gene Aliases
AT4G14965, AtMAPR4, membrane-associated progesterone binding protein 4
Chromosome Location
Chr.4: 8551244 - 8553618 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117583.8 NP_567451.1 5 - 6 457 80
GenBank mRNA AK229457.1 - 5 - 6 457 80
BT024469.1 - 5 - 6 345 80

View Full Product Details