Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACR4
Entrez Gene ID
843236
Gene Name
ACT domain repeat 4
Gene Aliases
ACT domain repeat 4, AT1G69040, F4N2.2, F4N2_2
Chromosome Location
Chr.1: 25957567 - 25960401 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105575.3 NP_177067.1 2 - 3 485 154
NM_202378.2 NP_974107.1 3 - 4 400 154
GenBank mRNA AF528060.1 - 2 - 3 146 154
BT029177.1 - 2 - 3 112 154

View Full Product Details