Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MIOX4
Entrez Gene ID
828732
Gene Name
myo-inositol oxygenase 4
Gene Aliases
AT4G26260, MYO-INOSITOL OXYGENASE 4, T25K17.70, T25K17_70, myo-inositol oxygenase 4
Chromosome Location
Chr.4: 13297695 - 13300444 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203915.2 NP_001190844.1 6 - 7 719 115
NM_118759.5 NP_194356.2 6 - 7 824 115
GenBank mRNA AY142501.1 - 6 - 7 580 115

View Full Product Details