Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRFL9
Entrez Gene ID
820436
Gene Name
TRF-like 9
Gene Aliases
AT3G12560, ATTBP2, TELOMERIC DNA-BINDING PROTEIN 2, TRF-like 9
Chromosome Location
Chr.3: 3981880 - 3985625 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338001.1 NP_001327482.1 3 - 4 1194 124
NM_112091.4 NP_187862.1 3 - 4 1157 124
GenBank mRNA AK227790.1 - 3 - 4 1082 124
AY550302.1 - 2 - 3 795 124
BT006452.1 - 2 - 3 795 124

View Full Product Details