Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HXK1
Entrez Gene ID
829034
Gene Name
hexokinase 1
Gene Aliases
ARABIDOPSIS THALIANA HEXOKINASE 1, AT4G29130, ATHXK1, F19B15.160, F19B15_160, GIN2, GLUCOSE INSENSITIVE 2, HEXOKINASE, hexokinase 1
Chromosome Location
Chr.4: 14352037 - 14355201 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119057.4 NP_194642.1 1 - 2 609 92
GenBank mRNA AK230265.1 - - 2436 92
AY075658.1 - 1 - 2 499 92
AY124809.1 - 1 - 2 273 92
U28214.1 - 1 - 2 470 92

View Full Product Details