Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SKB1
Entrez Gene ID
829240
Gene Name
SHK1 binding protein 1
Gene Aliases
AT4G31120, ATPRMT5, CAU1, F6E21.40, F6E21_40, PRMT5, PROTEIN ARGININE METHYLTRANSFERASE 5, SHK1 binding protein 1, calcium underaccumulation 1
Chromosome Location
Chr.4: 15132011 - 15136696 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119262.6 NP_194841.2 12 - 13 1161 78
NM_202918.2 NP_974647.1 12 - 13 1102 78

View Full Product Details