Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KNAT5
Entrez Gene ID
829335
Gene Name
homeobox protein knotted-1-like 5
Gene Aliases
AT4G32040, F10N7.150, F10N7_150, KNOTTED1-like homeobox gene 5, TRANSCRIPTION FACTOR
Chromosome Location
Chr.4: 15493989 - 15496381 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119356.4 NP_194932.1 5 - 6 1273 63
GenBank mRNA AF306661.1 - 5 - 6 1153 63
D43962.1 - 5 - 6 1179 63

View Full Product Details