Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
THIC
Entrez Gene ID
817513
Gene Name
thiaminC
Gene Aliases
AT2G29630, PY, PYRIMIDINE REQUIRING, T27A16.27, T27A16_27, thiaminC
Chromosome Location
Chr.2: 12666935 - 12671055 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202705.1 NP_001189634.1 2 - 3 491 138
NM_128517.4 NP_180524.1 2 - 3 447 138
NM_179804.3 NP_850135.1 2 - 3 447 138
GenBank mRNA AY092989.1 - 2 - 3 392 138
AY128756.1 - 1 - 2 300 138

View Full Product Details