Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sks15
Entrez Gene ID
829870
Gene Name
SKU5 similar 15
Gene Aliases
AP22.55, AP22_55, AT4G37160, SKU5 similar 15, SKU5 similar 15
Chromosome Location
Chr.4: 17494512 - 17497276 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342434.1 NP_001329607.1 6 - 7 1513 90
NM_119879.3 NP_195433.1 7 - 8 1580 90

View Full Product Details