Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UBP2
Entrez Gene ID
839397
Gene Name
ubiquitin-specific protease 2
Gene Aliases
AT1G04860, ATUBP2, F13M7.15, F13M7_15, UBIQUITIN-SPECIFIC PROTEASE 2, ubiquitin-specific protease 2
Chromosome Location
Chr.1: 1369036 - 1373103 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_100364.4 NP_563719.1 2 - 3 2927 74
GenBank mRNA AF302659.1 - 2 - 3 2879 74

View Full Product Details