Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TUB6
Entrez Gene ID
831100
Gene Name
beta-6 tubulin
Gene Aliases
AT5G12250, MXC9.21, MXC9_21, beta-6 tubulin
Chromosome Location
Chr.5: 3960938 - 3963624 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121263.4 NP_196786.1 2 - 3 627 67
GenBank mRNA AF360260.1 - 2 - 3 462 67
AY040074.1 - 1 - 2 390 67

View Full Product Details