Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BLH5
Entrez Gene ID
817264
Gene Name
BEL1-like homeodomain 5
Gene Aliases
AT2G27220, BEL1-like homeodomain 5, T22O13.1, T22O13_1
Chromosome Location
Chr.2: 11637106 - 11639809 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336110.1 NP_001318297.1 2 - 3 554 89
NM_128280.3 NP_180290.2 1 - 2 549 89
GenBank mRNA AB493565.1 - 1 - 2 435 89
DQ056543.1 - 1 - 2 435 89

View Full Product Details