Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LAG1
Entrez Gene ID
822140
Gene Name
TRAM, LAG1 and CLN8 (TLC) lipid-sensing domain containing protein
Gene Aliases
AT3G25540, LAG One Homologue 1, LAG1 HOMOLOG 1, LOH1, LONGEVITY ASSURANCE GENE 1
Chromosome Location
Chr.3: 9274335 - 9276685 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338755.1 NP_001319643.1 4 - 5 1040 90
NM_113450.5 NP_566769.1 5 - 6 1044 90
GenBank mRNA AF198179.1 - 4 - 5 769 90
AF360152.1 - 4 - 5 875 90
AY142597.1 - 4 - 5 719 90

View Full Product Details