Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HVA22A
Entrez Gene ID
843793
Gene Name
HVA22 homologue A
Gene Aliases
AT1G74520, ATHVA22A, F1M20.20, F1M20_20, HVA22 homologue A
Chromosome Location
Chr.1: 28007961 - 28009451 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001334652.1 NP_001323240.1 3 - 4 270 88
NM_106112.3 NP_177592.1 2 - 3 386 88
GenBank mRNA AF141659.1 - 2 - 3 144 88
AK118825.1 - 2 - 3 145 88
BT005492.1 - 2 - 3 91 88

View Full Product Details