Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TPS1
Entrez Gene ID
844194
Gene Name
trehalose-6-phosphate synthase
Gene Aliases
AT1G78580, ATTPS1, T30F21.9, T30F21_9, TREHALOSE-6-PHOSPHATE SYNTHASE 1, trehalose-6-phosphate synthase
Chromosome Location
Chr.1: 29552340 - 29560912 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001334832.1 NP_001319403.1 8 - 9 3250 78
NM_001334833.1 NP_001322931.1 7 - 8 3209 78
NM_001334834.1 NP_001322932.1 7 - 8 3176 78
NM_106505.5 NP_177979.1 7 - 8 1440 78
GenBank mRNA KJ138926.1 - 6 - 7 1234 78

View Full Product Details