Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RST1
Entrez Gene ID
822389
Gene Name
ARM repeat superfamily protein
Gene Aliases
AT3G27670, RESURRECTION1
Chromosome Location
Chr.3: 10245112 - 10253308 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_113683.3 NP_189404.2 19 - 20 5274 97

View Full Product Details