Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HMG
Entrez Gene ID
822505
Gene Name
high mobility group
Gene Aliases
AT3G28730, ATHMG, NFD, NUCLEOSOME/CHROMATIN ASSEMBLY FACTOR D, SSRP1, high mobility group
Chromosome Location
Chr.3: 10784888 - 10788729 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_113794.4 NP_189515.1 8 - 9 924 122
GenBank mRNA BT002507.1 - 8 - 9 918 122

View Full Product Details