Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KIN11
Entrez Gene ID
822566
Gene Name
SNF1 kinase homolog 11
Gene Aliases
AKIN11, AT3G29160, ATKIN11, SNF1 kinase homolog 11, SNF1-RELATED PROTEIN KINASE, SNF1-RELATED PROTEIN KINASE 1.2, SNRK1.2
Chromosome Location
Chr.3: 11128620 - 11132105 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_113839.4 NP_566843.1 10 - 11 1730 96
NM_202645.3 NP_974374.1 10 - 11 1582 96
NM_202646.1 NP_974375.1 10 - 11 1503 96
GenBank mRNA AY070468.1 - 10 - 11 1533 96
AY149927.1 - 9 - 10 1430 96
DQ778956.1 - 9 - 10 1430 96
X99279.1 - - 1516 96

Gene Symbol
AT2G15960
Entrez Gene ID
816091
Gene Name
stress-induced protein
Gene Aliases
AT2G15960, F19G14.4, F19G14_4
Chromosome Location
Chr.2: 6947096 - 6947866 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK230337.1 - - 215 96

View Full Product Details