Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATSMC2
Entrez Gene ID
823900
Gene Name
Structural maintenance of chromosomes (SMC) family protein
Gene Aliases
AT3G47460
Chromosome Location
Chr.3: 17486490 - 17493395 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339310.1 NP_001325474.1 10 - 11 2372 94
NM_114614.4 NP_190330.1 10 - 11 2309 94

View Full Product Details