Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SHM6
Entrez Gene ID
838807
Gene Name
serine hydroxymethyltransferase 6
Gene Aliases
AT1G22020, F2E2.7, F2E2_7, serine hydroxymethyltransferase 6
Chromosome Location
Chr.1: 7754281 - 7758029 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332533.1 NP_001323098.1 3 - 4 1896 118
NM_102052.3 NP_173621.1 3 - 4 1873 118
GenBank mRNA AY125514.1 - 3 - 4 1856 118
BT004532.1 - 3 - 4 1578 118

View Full Product Details