Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SQP2
- Entrez Gene ID
-
832479
- Gene Name
- squalene monooxygenase 2
- Gene Aliases
-
AT5G24140, MLE8.6, MLE8_6, squalene monooxygenase 2
- Chromosome Location
-
Chr.5: 8164319 - 8167776 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_122319.4 |
NP_197802.2 |
1 - 2 |
304 |
76 |
View Full Product Details