Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CLC-A
Entrez Gene ID
834090
Gene Name
chloride channel A
Gene Aliases
AT5G40890, ATCLC-A, ATCLCA, CHLORIDE CHANNEL A, CHLORIDE CHANNEL-A, CLCA, chloride channel A
Chromosome Location
Chr.5: 16381314 - 16385424 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036913.2 NP_001031990.1 3 - 4 1283 104
NM_123454.3 NP_198905.1 4 - 5 1513 104
GenBank mRNA AF044313.1 - 4 - 5 1334 104
AY150506.1 - 4 - 5 1088 104

View Full Product Details