Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IPK1
Entrez Gene ID
834292
Gene Name
inositol-pentakisphosphate 2-kinase 1
Gene Aliases
AT5G42810, ATIPK1, M40H3, MJB21.19, MJB21_19, inositol-pentakisphosphate 2-kinase 1
Chromosome Location
Chr.5: 17166702 - 17169459 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_123646.4 NP_568613.1 5 - 6 993 151
GenBank mRNA AY059898.1 - 5 - 6 868 151
AY093362.1 - 5 - 6 622 151
DQ122931.1 - 5 - 6 622 151

View Full Product Details