The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_129647.4 | NP_181616.3 | 4 - 5 | 789 | 108 |
GenBank mRNA | AY081315.1 | - | 4 - 5 | 580 | 108 |
BT010364.1 | - | 3 - 4 | 531 | 108 |