Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WRKY56
Entrez Gene ID
842703
Gene Name
WRKY DNA-binding protein 56
Gene Aliases
AT1G64000, ATWRKY56, F22C12.23, F22C12_23, WRKY DNA-binding protein 56
Chromosome Location
Chr.1: 23747304 - 23749053 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105073.2 NP_176583.1 1 - 2 528 154
GenBank mRNA AY071848.1 - 1 - 2 398 154
BT024835.1 - 1 - 2 398 154

View Full Product Details