Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABCB13
Entrez Gene ID
839687
Gene Name
P-glycoprotein 13
Gene Aliases
AT1G27940, ATP-binding cassette B13, F13K9.5, F13K9_5, P-glycoprotein 13, PGP13
Chromosome Location
Chr.1: 9733430 - 9738236 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332792.1 NP_001322427.1 2 - 3 437 176
NM_102559.2 NP_174115.1 2 - 3 437 176

View Full Product Details