Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UGD1
Entrez Gene ID
839197
Gene Name
UDP-glucose dehydrogenase 1
Gene Aliases
AT1G26570, ATUGD1, T1K7.6, T1K7_6, UDP-GLUCOSE DEHYDROGENASE 1, UDP-glucose dehydrogenase 1
Chromosome Location
Chr.1: 9182008 - 9184393 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_102419.4 NP_173979.1 2 - 2 1848 120
GenBank mRNA AY070758.1 - 2 - 2 1651 120

View Full Product Details